Skip to Content
Merck
All Photos(1)

Key Documents

47742

Supelco

Folin-Denis′ reagent

for the determination of phenols

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

quality

for the determination of phenols

specific analyte(s)

phenol

technique(s)

photometry: suitable

refractive index

n20/D 1.352

application(s)

food and beverages
general analytical

Looking for similar products? Visit Product Comparison Guide

General description

Folin-Denis′ reagent can be prepared by mixing mixture of sodium tungstate and (phospho)molybdic acid in phosphoric acid. This reagent is generally used in determination of phenolics in urine.
Folin-Denis′ reagent has been used for the determination of total phenolic compounds present in the peanut hulls spectrophotometrically.

Application

Folin-Denis′ reagent has been used in the determination of total phenolics in the fruits of Silybum marianum using spectrophotometry technique.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Giusti et al.
International journal of food sciences and nutrition, 69(5), 557-565 (2017-11-10)
Consumption of legumes has become popular among health-conscious consumers due to the high levels of nutrients such as proteins, dietary fibres, minerals, vitamins and micronutrients like polyphenols. Ten legumes cultivars organically and conventionally grown were compared regarding their chemical profiles
Phenolic Compound Biochemistry
Wilfred Vermerris, Ralph Nicholson
Springer Science & Business Media, 152-152 (2007)
Scavenging effect of methanolic extracts of peanut hulls on free-radical and active-oxygen species
Yen, Gow Chin, and Pin Der Duh.
Journal of Agricultural and Food Chemistry, 42 (3), 629-632 (1994)
Claudia Giuliani et al.
Fitoterapia, 130, 210-218 (2018-09-15)
Silybum marianum (L.) Gaertn (Asteraceae) is a valuable medicinal plant utilized for silymarin production. However, only fragmentary and contradictory information about silymarin localization within S. marianum fruit are available. In this work, a twofold research approach was adopted in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service