Skip to Content
Merck
All Photos(1)

Key Documents

46995

Supelco

Xylazine hydrochloride

VETRANAL®, analytical standard

Synonym(s):

2-(2,6-Dimethylphenylamino)-5,6-dihydro-4H-thiazine hydrochloride, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2S · HCl
CAS Number:
Molecular Weight:
256.79
Beilstein:
6448471
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical
forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Cc1cccc(C)c1NC2=NCCCS2

InChI

1S/C12H16N2S.ClH/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12;/h3,5-6H,4,7-8H2,1-2H3,(H,13,14);1H

InChI key

QYEFBJRXKKSABU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

α2-Adrenergic receptor agonist, sedative, muscle relaxant.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henry P Farhoodi et al.
Journal of bone oncology, 23, 100298-100298 (2020-07-10)
Many cancers metastasize to the bones, particularly in cases of breast and prostate cancers. Due to the "vicious cycle" of cancer cells inducing bone resorption, which promotes further tumor growth, they are difficult to treat and may lead to extreme
Ori Furman et al.
Theriogenology, 82(8), 1165-1172 (2014-09-13)
This study establishes an experimental model for subclinical mastitis induced by Gram-positive (G+) exosecretions of Staphylococcus aureus origin or Gram-negative (G-) endotoxin of Escherichia coli origin to examine its effects on follicular growth and steroid concentrations in Holstein dairy cows.
Javier Gutiérrez-Cuesta et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(13), 2974-2988 (2014-06-20)
The repeated cycles of cessation of consumption and relapse remain the major clinical concern in treating drug addiction. The endogenous opioid system is a crucial component of the reward circuit that participates in the adaptive changes leading to relapse in
Burak Turgut et al.
Clinical & experimental ophthalmology, 43(1), 67-76 (2014-05-08)
To investigate the effects of subconjunctivally administered trastuzumab on wound healing in experimental glaucoma filtration surgery. Comparative, experimental study. Twenty eight eyes. Twenty-eight male New Zealand white rabbits were randomly assigned to four groups, each including seven rabbits: The rabbits
Marzia Ferretti et al.
Histology and histopathology, 29(8), 1027-1037 (2014-02-11)
The present paper completes our recent study on the effects of phytoestrogen ferutinin in preventing osteoporosis and demonstrating the superior osteoprotective effect of a 2 mg/kg/day dose in ovariectomized (OVX) rats, compared to both estrogens and lower (0.5, 1 mg/kg/day)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service