Skip to Content
Merck
All Photos(3)

Key Documents

46337

Supelco

17α-Hydroxyprogesterone

VETRANAL®, analytical standard

Synonym(s):

17α-Hydroxy-4-pregnene-3,20-dione, 4-Pregnen-17α-ol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
Beilstein:
3218109
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(C)=O

InChI

1S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1

InChI key

DBPWSSGDRRHUNT-CEGNMAFCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Jeong Kim et al.
Journal of Korean medical science, 27(11), 1439-1443 (2012-11-21)
Congenital adrenal hyperplasia (CAH) is characterized by decreased adrenal hormone production due to enzymatic defects and subsequent rise of adrenocorticotrophic hormone that stimulates the adrenal cortex to become hyperplastic, and sometimes tumorous. As the pathophysiology is basically a defect in
O Lekarev et al.
Journal of perinatology : official journal of the California Perinatal Association, 33(1), 76-78 (2012-12-28)
Congenital adrenal hyperplasia (CAH) is an autosomal recessive disorder where steroidogenesis in the adrenal cortex is impaired. The most common form is caused by 21-hydroxylase deficiency (21OHD). Classical 21OHD is characterized by glucocorticoid and mineralocorticoid deficiency and by overproduction of
Adriana Pabón et al.
Molecules (Basel, Switzerland), 18(3), 3356-3378 (2013-03-16)
Solanum nudum Dunal steroids have been reported as being antimalarial compounds; however, their concentration in plants is low, meaning that the species could be threatened by over-harvesting for this purpose. Swern oxidation was used for hemisynthesis of diosgenone (one of
F De Amicis et al.
Animal reproduction science, 135(1-4), 75-84 (2012-10-11)
Progesterone receptors (PR) through interaction with the specific ligand progesterone (PRG), play a central coordinate role in different reproductive events. In this study conventional PR were identified in boar spermatozoa by Western blotting. Immunofluorescence techniques indicate that PR are located
Stefan Németh et al.
Clinica chimica acta; international journal of clinical chemistry, 414, 211-214 (2012-09-19)
Congenital adrenal hyperplasia (CAH) is an autosomal recessive disorder most commonly caused by defects in the CYP21A2 gene. Neonatal CAH-screening based on 17-hydroxyprogesterone (17-OHP) measurements prevents life-threatening salt wasting conditions in newborns, but results in a considerable false-positive rate. Therefore

Articles

Milli-Q® ultrapure water systems deliver water free of organic contaminants , including hormones present in a lab's tap water, making it suitable for the most sensitive LC-MS/MS analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service