Skip to Content
Merck

Skip To

46331

Tepraloxydim

PESTANAL®, analytical standard

Synonym(s):

trans-2-[1-(3-Chloroallyloxyimino)propyl]-3-hydroxy-5-(tetrahydro-2H-pyran-4-yl)-2-cyclohexen-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
$163.00

About This Item

Empirical Formula (Hill Notation):
C17H24ClNO4
CAS Number:
Molecular Weight:
341.83
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC\C(=N/OC\C=C\Cl)C1=C(O)CC(CC1=O)C2CCOCC2

InChI

1S/C17H24ClNO4/c1-2-14(19-23-7-3-6-18)17-15(20)10-13(11-16(17)21)12-4-8-22-9-5-12/h3,6,12-13,20H,2,4-5,7-11H2,1H3/b6-3+,19-14+

InChI key

IOYNQIMAUDJVEI-BMVIKAAMSA-N

General description

Tepraloxydim belongs to cyclohexanedione class. It is a post-emergence herbicide, which inhibits the enzyme activity of acetyl-CoA carboxylase involved in the catalysis of fatty acid synthesis.

Application

Tepraloxydim may be used as a reference standard for the determination of tepraloxydim in drinking water by solid-phase extraction and liquid chromatography with UV detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
759743167635346
format

neat

format

neat

format

neat

format

neat

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

analytical standard

grade

analytical standard

product line

PESTANAL®

product line

TraceCERT®

product line

PESTANAL®

product line

PESTANAL®

Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

100

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?

Explore all of our products under Tepraloxydim


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters