Skip to Content
Merck
All Photos(1)

Key Documents

46263

Supelco

17α-Ethynylestradiol

VETRANAL®, analytical standard

Synonym(s):

17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
2419975
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

182-183 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@@]3(O)C#C)[C@]1([H])CCc4cc(O)ccc24

InChI

1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1

InChI key

BFPYWIDHMRZLRN-SLHNCBLASA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Han et al.
Water research, 47(7), 2273-2284 (2013-03-08)
This study demonstrates that ethinylestradiol (EE2), a priority estrogenic contaminant in water, can be rapidly and selectively removed from aqueous solutions using industrial-grade polyamide 612 (PA612) particles as adsorbents. Isothermal studies showed that nonporous low surface area (20 m(2) g(-1)) PA612 particles
Bernardine H Stegeman et al.
BMJ (Clinical research ed.), 347, f5298-f5298 (2013-09-14)
To provide a comprehensive overview of the risk of venous thrombosis in women using different combined oral contraceptives. Systematic review and network meta-analysis. PubMed, Embase, Web of Science, Cochrane, Cumulative Index to Nursing and Allied Health Literature, Academic Search Premier
Konrad F Koehler et al.
Endocrine reviews, 26(3), 465-478 (2005-04-29)
We have known for many years that estrogen is more than the female hormone. It is essential in the male gonads, and in both sexes, estrogen has functions in the skeleton and central nervous system, on behavior, and in the
Lourdes Ibáñez et al.
The Journal of clinical endocrinology and metabolism, 98(5), E902-E907 (2013-04-03)
An oral estro-progestagen is the standard medication given to adolescent girls with androgen excess, even when those girls are not at risk of pregnancy. The aim of this study was to compare on-treatment and post-treatment effects of intervention with an
Laura Moro et al.
Blood, 105(1), 115-121 (2004-06-17)
The impact of estrogens on the cardiovascular system and their ability to regulate platelet function are matters of controversy. The recent finding that estrogen receptors are expressed in human platelets renders these cells an excellent model for studying the nongenomic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service