Skip to Content
Merck
All Photos(1)

Documents

46028

Supelco

Thifensulfuron-methyl

PESTANAL®, analytical standard

Synonym(s):

Harmony

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N5O6S2
CAS Number:
Molecular Weight:
387.39
Beilstein:
7448062
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1sccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2

InChI

1S/C12H13N5O6S2/c1-6-13-10(16-12(14-6)23-3)15-11(19)17-25(20,21)7-4-5-24-8(7)9(18)22-2/h4-5H,1-3H3,(H2,13,14,15,16,17,19)

InChI key

AHTPATJNIAFOLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thifensulfuron-methyl belongs to the class of sulfonylurea pesticides, generally employed for controlling many grasses and broadleaved weed species, affecting cultivation of crops such as corn, potatoes, tomatoes, citrus, vines and rice.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thifensulfuron-methyl may be used as an analytical reference standard for the quantification of the analyte in environmental matrices using various chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of sulfonylurea degradation products in soil by liquid chromatography-ultraviolet detection followed by confirmatory liquid chromatography-tandem mass spectrometry
Bossi R, et al.
Journal of Chromatography A, 855(2), 575-582 (1999)
Christine A Riedy et al.
BMC oral health, 8, 5-5 (2008-02-13)
Xylitol chewing gum has been shown to reduce Streptococcus mutans levels and decay. Two studies examined the presence and time course of salivary xylitol concentrations delivered via xylitol-containing pellet gum and compared them to other xylitol-containing products. A within-subjects design
Qing-Zhi Zhu et al.
Environmental science & technology, 36(24), 5411-5420 (2003-01-11)
A molecularly imprinted polymer (MIP) was synthesized using the herbicide metsulfuron-methyl (MSM) as a template, 2-(trifluoromethyl)acrylic acid as a functional monomer, divinylbenzene as a cross-linker, and dichloromethane as a porogen. This polymer was used as a solid-phase extraction material for
Allan J Cessna et al.
Journal of environmental quality, 35(6), 2395-2401 (2006-10-31)
Sulfonylurea herbicides are applied at relatively low rates (3 to 40 g ha(-1)) to control weeds in a variety of crops across the Canadian prairies. Because of their high phytotoxicity and the likelihood of their transport in surface runoff, there
Cheng-gen Xie et al.
Luminescence : the journal of biological and chemical luminescence, 26(4), 271-279 (2010-07-16)
Uniform molecular imprinting microspheres were prepared using precipitation polymerization with thifensulfuron-methyl (TFM) as template, acrylamide as functional monomer and ethylene glycol dimethacrylate as cross-linker. TFM could be selectively adsorbed on the molecularly imprinted polymers (MIPs) matrix through the hydrogen bonding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service