Skip to Content
Merck
All Photos(1)

Key Documents

45713

Supelco

TBTC

PESTANAL®, analytical standard

Synonym(s):

Tributyltin chloride, TBTC, Tributylchlorotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnCl
CAS Number:
Molecular Weight:
325.51
Beilstein:
3535715
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.492 (lit.)

bp

171-173 °C/25 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCC[Sn](Cl)(CCCC)CCCC

InChI

1S/3C4H9.ClH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1

InChI key

GCTFWCDSFPMHHS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eiji Munetsuna et al.
Endocrine research, 39(4), 168-172 (2014-04-01)
Hippocampal functions are influenced by steroid hormones, such as testosterone and estradiol. It has been demonstrated that hippocampus-derived steroid hormones play important roles in neuronal protection and synapse formation. Our research groups have demonstrated that estradiol is de novo synthesized
Outi Huttala et al.
ALTEX, 36(3), 419-434 (2019-03-18)
Besides being an energy storage, adipose tissue is an endocrine organ closely associated with vascular system. Human relevant in vitro models are needed to study adipose tissue and related diseases. Vasculature plays a central role in the development and inhibition
Flaviana T Vieira et al.
European journal of medicinal chemistry, 45(3), 883-889 (2009-12-05)
The reaction of 1,3-cyclohexadione with 2-aminobenzoic acid has produced the 2-(3-oxocyclohex-1-enyl)benzoic acid (HOBz). Subsequent reactions of the ligand with organotin chlorides led to [Me(2)Sn(OBz)O](2) (1), [Bu(2)Sn(OBz)O](2) (2), [Ph(2)Sn(OBz)O](2) (3), [Me(3)Sn(OBz)] (4), [Bu(3)Sn(OBz)] (5) and [Ph(3)Sn(OBz)] (6). All complexes have been
Tijana Markovič et al.
Toxicology letters, 233(1), 8-15 (2014-12-30)
Evaluating immunomodulatory effects of xenobiotics is an important component of the toxicity studies. Herein we report on the establishment of a novel invitro test system for the immunotoxicity screening of xenobiotics based on human lymphoblastoid cell lines (LCLs). Four immunotoxic
Sumonto Mitra et al.
Environmental toxicology and pharmacology, 37(3), 1048-1059 (2014-04-26)
Tributyltin (TBT) is a potent biocide and commonly used in various industrial sectors. Humans are mainly exposed through the food chain. We have previously demonstrated tin accumulation in brain following TBT-chloride (TBTC) exposure. In this study, effect of TBTC on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service