Skip to Content
Merck
All Photos(2)

Key Documents

45665

Supelco

Sulprofos

PESTANAL®, analytical standard

Synonym(s):

Sulprophos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19O2PS3
CAS Number:
Molecular Weight:
322.45
Beilstein:
1990231
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCSP(=S)(OCC)Oc1ccc(SC)cc1

InChI

1S/C12H19O2PS3/c1-4-10-18-15(16,13-5-2)14-11-6-8-12(17-3)9-7-11/h6-9H,4-5,10H2,1-3H3

InChI key

JXHJNEJVUNHLKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulprofos is a selective organophosphorus insecticide, primarily used in cotton cultivation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulprofos may be used as an analytical reference standard for the quantification of the analyte in Radix astragali root and fruit juice samples using different chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic Pathways of Agrochemicals: Part 2: Insecticides and Fungicides (2007)
Application of magnetic solvent bar liquid-phase microextraction for determination of organophosphorus pesticides in fruit juice samples by gas chromatography mass spectrometry
Wu L, et al.
Food Chemistry, 176(10), 197-204 (2015)
Toxicology and Pesticide Use in Relation to Wildlife, Organophosphorus, and Carbamate Compounds (1992)
Simultaneous determination of organophosphorus, organochlorine, pyrethriod and carbamate pesticides in Radix astragali by microwave-assisted extraction/dispersive-solid phase extraction coupled with GC-MS
Mao X, et al.
Talanta, 97(10), 131-141 (2012)
Frank J Byrne et al.
Journal of economic entomology, 95(2), 425-429 (2002-05-22)
Peracid activated organophosphorus (OP) insecticides were evaluated against electric eel and a range of insect acetylcholinesterases (AChE). Incubation of chlorpyrifos and sulprofos with meta-chloroperoxybenzoic acid (MCPBA) produced active metabolites capable of distinguishing between AChE enzymes conferring OP susceptibility and resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service