Skip to Content
Merck
All Photos(1)

Key Documents

45570

Supelco

Metam-sodium hydrate

PESTANAL®, analytical standard

Synonym(s):

Sodium methyldithiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4NNaS2 · xH2O
Molecular Weight:
129.18 (anhydrous basis)
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

CNC(S[Na])=S.[H]O[H]

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

M Villarino et al.
Plant disease, 103(8), 1974-1982 (2019-06-19)
Fusarium solani is an emerging pathogen reported on Spanish strawberry crops both in nurseries and in fruit production fields, causing wilt and root rot. Pathogenicity, morphocultural characteristics, and sensitivity to biocides of 103 F. solani isolates recovered from symptomatic strawberry
Lauren Gay et al.
Avian diseases, 54(1 Suppl), 763-767 (2010-06-05)
The chemical compound metam-sodium was tested at three concentrations for the ability to inactivate the infectivity of low-pathogenic avian influenza virus (LPAIV) and infectious bursal disease virus (IBDV), with virus-contaminated chicken litter used as the substrate. LPAIV was inactivated within
M Díaz-Pérez et al.
Microbial ecology, 57(2), 379-383 (2008-11-11)
The monoculture of melon in Guatemala has caused the massive appearance of plants with an analogous syndrome for the well-known disease commonly called melon collapse, or vine decline, causing significant losses in crops. Methyl bromide is commonly used to sterilize
W T Runia et al.
Communications in agricultural and applied biological sciences, 72(3), 687-691 (2008-04-11)
In The Netherlands the only chemical alternative for methyl bromide permitted is an application of metam-sodium (MS) with the active ingredient methyl isothiocyanate (MIT). After introduction of a new application method with 'rotary spading injection' legislation restricted the application of
Stephen B Pruett et al.
Toxicological sciences : an official journal of the Society of Toxicology, 109(2), 237-246 (2009-04-03)
Sodium methyldithiocarbamate (SMD) is the third most abundantly used conventional pesticide in the United States, and hundreds of thousands of persons are exposed to this compound or its major breakdown product, methylisothiocyanate, at levels greater than recommended by the Environmental

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service