Skip to Content
Merck
All Photos(2)

Key Documents

45541

Supelco

Isoprocarb

PESTANAL®, analytical standard

Synonym(s):

2-Isopropylphenyl N-methylcarbamate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$109.00

$109.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$109.00

About This Item

Empirical Formula (Hill Notation):
C11H15NO2
CAS Number:
Molecular Weight:
193.24
Beilstein:
1875020
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$109.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1ccccc1C(C)C

InChI

1S/C11H15NO2/c1-8(2)9-6-4-5-7-10(9)14-11(13)12-3/h4-8H,1-3H3,(H,12,13)

InChI key

QBSJMKIUCUGGNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Xue Jin et al.
Journal of economic entomology, 110(2), 641-650 (2017-03-24)
Sogatella furcifera (Horváth) is a migratory insect that is one of the most important pest species on rice in many Asian countries. Control of S. furcifera (Hemiptera: Delphacidae) primarily depends on the use of chemical insecticides, and with this extensive
Flavio Della Pelle et al.
Talanta, 186, 389-396 (2018-05-23)
An electrochemical screening assay for the detection of phenyl carbamates (i.e. carbaryl, carbofuran, isoprocarb and fenobucarb) was developed and applied to grains samples (i.e. durum wheat, soft wheat and maize). Nano carbon black (CB) was strategically employed to realize an
Xiaolei Zhang et al.
Pesticide biochemistry and physiology, 132, 13-20 (2016-08-16)
The brown planthopper is a serious rice pest in China. Chemical insecticides have been considered a satisfactory means of controlling the brown planthopper. In the present study, we determined the susceptibility of twenty-one populations of Nilaparvata lugens to eleven insecticides
Pattamaporn Klongklaew et al.
Analytical and bioanalytical chemistry, 410(27), 7185-7193 (2018-09-01)
A hierarchically porous structured composite monolith sorbent of polypyrrole-coated graphene oxide and octadecyl silica incorporated in chitosan cryogel (PPY/GOx/C18/chitosan) was synthesized and used as solid-phase extraction sorbent for the determination of carbamate pesticides. Various factors affecting the characteristics of the
Chao Wang et al.
Environmental toxicology, 27(1), 50-57 (2010-06-16)
Organophosphates and carbamates are widely used pesticides and play an important role in global agriculture. The misuse of these compounds has caused environmental problems and has had a negative impact on wildlife. In this study, the in vivo effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service