Skip to Content
Merck
All Photos(1)

Key Documents

45514

Supelco

Formetanate hydrochloride

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N3O2 · HCl
CAS Number:
Molecular Weight:
257.72
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cl.CNC(=O)Oc1cccc(c1)\N=C\N(C)C

InChI

1S/C11H15N3O2.ClH/c1-12-11(15)16-10-6-4-5-9(7-10)13-8-14(2)3;/h4-8H,1-3H3,(H,12,15);1H/b13-8+;

InChI key

MYPKGPZHHQEODQ-FNXZNAJJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

store in bottles protecting against light.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynda V Podhorniak et al.
Journal of agricultural and food chemistry, 58(10), 5862-5867 (2010-04-17)
A rapid multiresidue method that captures residues of the insecticide formetanate hydrochloride (FHCl) in selected fruits is described. The method was used to provide residue data for dietary exposure determinations of FHCl. Using an acetonitrile extraction with a dispersive cleanup
Burak Polat et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 55(1), 1-10 (2019-09-17)
The effects of washing treatments on removal rates of some pesticides residues (acetamiprid, chlorpyrifos and formetanate hydrochloride) on pepper were investigated. Method verification was conducted through spiking pepper samples at 0.1, 1.0 and 10.0 × MRL. QuEChERS method produced average
V C Moser et al.
Pharmacology, biochemistry, and behavior, 26(1), 119-121 (1987-01-01)
Formetanate (FMT) is a formamidine acaricide/insecticide with a carbamate moiety in its molecular structure. FMT-induced lethality is reportedly due to inhibition of acetylcholinesterase. Here we report evidence of the neurochemical basis for the sublethal, behavioral effects of FMT in rats.
Martin Staroň et al.
Environmental science and pollution research international, 24(16), 14060-14070 (2017-04-15)
Substantial percentage of world food production depends on pollinating service of honeybees that directly depends on their health status. Among other factors, the success of bee colonies depends on health of developed larvae. The crucial phase of larval development is
Determination of the insecticide/acaricide formetanate in fresh fruit by reversed-phase liquid chromatography.
J F Lawrence et al.
Journal of agricultural and food chemistry, 29(4), 722-724 (1981-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service