Skip to Content
Merck
All Photos(1)

Documents

45446

Supelco

Diflubenzuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9ClF2N2O2
CAS Number:
Molecular Weight:
310.68
Beilstein:
2162461
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1cccc(F)c1C(=O)NC(=O)Nc2ccc(Cl)cc2

InChI

1S/C14H9ClF2N2O2/c15-8-4-6-9(7-5-8)18-14(21)19-13(20)12-10(16)2-1-3-11(12)17/h1-7H,(H2,18,19,20,21)

InChI key

QQQYTWIFVNKMRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J D Quigley et al.
Journal of dairy science, 102(3), 2242-2253 (2019-01-30)
Apparent total-tract digestibility data from 3 published studies with calves from 0 to 4 mo of age were used to evaluate National Research Council (NRC; 2001) estimates of digestible energy (DE) and metabolizable energy (ME) in calf starters (CS). Calves
Priscila Fernandes Viana-Medeiros et al.
PLoS neglected tropical diseases, 12(8), e0006734-e0006734 (2018-08-21)
Resistance to pyrethroids and to the organophosphate temephos is widespread in Brazilian populations of the dengue vector, Aedes aegypti. Thereof, since 2009 Insect Growth Regulators are employed as larvicides, and malathion is used against adults. We performed laboratory selection with
Annie J Krueger et al.
Environmental entomology, 50(5), 1105-1117 (2021-07-20)
Risk assessment for chemicals in the United States relies upon the honey bee (Apis meliffera L. [Hymenoptera: Apidae]) as a surrogate for other bee species. There is uncertainty in extrapolating honey bee toxicity data to bumble bees due to differences
Tobias Suter et al.
Parasites & vectors, 10(1), 431-431 (2017-09-21)
Aedes aegypti and Ae. albopictus are two highly invasive mosquito species, both vectors of several viruses, including dengue, chikungunya and Zika. While Ae. aegypti is the primary vector in the tropics and sub-tropics, Ae. albopictus is increasingly under the public
Thomas Van Leeuwen et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(12), 4407-4412 (2012-03-07)
Because of its importance to the arthropod exoskeleton, chitin biogenesis is an attractive target for pest control. This point is demonstrated by the economically important benzoylurea compounds that are in wide use as highly specific agents to control insect populations.

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service