Skip to Content
Merck
All Photos(1)

Key Documents

45439

Supelco

1,3-Dichloropropane

PESTANAL®, analytical standard

Synonym(s):

Trimethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3Cl
CAS Number:
Molecular Weight:
112.99
Beilstein:
505960
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.448 (lit.)

bp

120-122 °C (lit.)

mp

−99 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

ClCCCCl

InChI

1S/C3H6Cl2/c4-2-1-3-5/h1-3H2

InChI key

YHRUOJUYPBUZOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Rodriguez-Arnaiz
Environmental and molecular mutagenesis, 31(4), 390-401 (1998-07-08)
Biotransformation of several structurally related 2B compounds to reactive metabolites was evaluated in the somatic w/w+ assay of Drosophila melanogaster. Chemicals tested were the dichlorinated alkanes dichloromethane (DCM), 1,2-dichloroethane (DCE), and 1,3-dichloropropane (DCP); the thiouracil derivatives 5-methyl, 2-thiouracil (5M2TU), 6-methyl
1,3-Dichloropropene (technical grade).
Report on carcinogens : carcinogen profiles, 10, 91-93 (2004-08-25)
Stefaan De Neve et al.
Journal of environmental quality, 33(5), 1647-1652 (2004-09-10)
The effect of soil fumigation on N mineralization and nitrification needs to be better quantified to optimize N fertilizer advice and predict NO(-)(3) concentrations in crops and NO(-)(3) leaching risks. Seven soils representing a range in soil texture and organic
D W Herr et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 35(1), 31-48 (1997-01-01)
Previous research showed that acute exposure to dichloromethane (DCM) produced a selective reduction in peak N30 of flash evoked potentials (FEPs) in rats. In contrast, acute exposures to p-xylene or toluene selectively reduced FEP peak N160. The present experiments compared
Biological effect monitoring.
C T Evelo et al.
Archives of toxicology. Supplement. = Archiv fur Toxikologie. Supplement, 15, 268-277 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service