Skip to Content
Merck
All Photos(1)

Documents

45395

Supelco

Chlorpyrifos

PESTANAL®, analytical standard

Synonym(s):

Chlorpyriphos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11Cl3NO3PS
CAS Number:
Molecular Weight:
350.59
Beilstein:
1545756
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3

InChI key

SBPBAQFWLVIOKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorpyrifos belongs to the organophosphorus class of insecticides, effectively used for the control of a variety of insect pests such as mosquitoes, fire ants, cockroaches, horn flies, lice, fleas, turf and ornamental plant insects.

Application

Chlorpyrifos may be used as an analytical reference standard for the quantification of the analyte in waste discharges and biological samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Eisler's Encyclopedia of Environmentally Hazardous Priority Chemicals (2007)
Validation of UV Spectrophotometric and HPLC Methods for Quantitative determination of chlorpyrifos
Zalat.A.O, et al.
International Letters of Chemistry, Physics and Astronomy, 2, 58-63 (2014)
Shaza Gaballah et al.
Environmental health perspectives, 128(4), 47005-47005 (2020-04-10)
Per- and polyfluoroalkyl substances (PFAS) are a diverse class of industrial chemicals with widespread environmental occurrence. Exposure to long-chain PFAS is associated with developmental toxicity, prompting their replacement with short-chain and fluoroether compounds. There is growing public concern over the
Liquid chromatography--tandem mass spectrometry method for the simultaneous quantitative determination of the organophosphorus pesticides dimethoate, fenthion, diazinon and chlorpyrifos in human blood
Salm P, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 877(5-6), 568-574 (2009)
D J Clegg et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 2(3), 257-279 (1999-08-03)
A panel of toxicology and medical experts was convened on 7-9 April 1997 to consider the available scientific literature on chlorpyrifos, both published and unpublished, to determine the acute and chronic toxicology reference dose (RfD). In the course of reviewing

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service