Skip to Content
Merck
All Photos(1)

Key Documents

44689

Supelco

Kuromanin chloride

analytical standard

Synonym(s):

Asterin, Chrysanthemin, Cyanidin 3-O-glucoside chloride, Glucocyanidin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2=CC3=C([O+]=C2C4=CC(O)=C(O)C=C4)C=C(O)C=C3O)O[C@H](CO)[C@H]1O.[Cl-]

InChI

1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17-,18+,19-,21-;/m1./s1

InChI key

YTMNONATNXDQJF-UBNZBFALSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anthocyanin. Antioxidant. Studied for its neuroprotective property.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacynthe Desjardins et al.
Journal of medicinal food, 15(12), 1045-1050 (2012-06-29)
Periodontal diseases are a group of multifactorial polymicrobial infections characterized by a progressive inflammatory destruction of the periodontium. Flavonoids, including anthocyanins, are receiving increasing attention because of their promising human health benefits. The aim of our study was to investigate
Yun Wang et al.
The Journal of nutrition, 142(6), 1033-1037 (2012-04-27)
In this study, we investigated the protective effects of the anthocyanin cyanidin-3-O-β-glucoside (C3G) on hypercholesterolemia-induced endothelial dysfunction in apoE-deficient (apoE(-/-)) mice. In the prevention study, twenty 8-wk-old male apoE(-/-) mice (n = 10/group) were fed a high-fat, cholesterol-rich diet (HCD)
Jesús Aguayo-Rojas et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 67(2), 178-185 (2012-05-09)
The lime-cooking extrusion represents an alternative technology for manufacturing pre-gelatinized flours for tortillas with the advantages of saving energy and generation of null effluents. The phytochemical profiles (total phenolics, anthocyanins) and antioxidant activity of four different types of whole pigmented
Zhou Zhou et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(10), 878-881 (2012-06-19)
Studies have shown positive correlations between inflammatory cytokines such as interleukin-6 (IL-6) and the development of chronic diseases including cardiovascular disease by activating C-reactive protein (CRP). Both atorvastatin calcium (lipitor) as well as flavonoid rich fruit such as tart cherry
Alzbeta Kamenickova et al.
Toxicology letters, 221(1), 1-8 (2013-06-06)
Anthocyanins are plant pigments occurring in flowers and berry fruits. Since a phenomenon of food-drug interactions is increasingly emerging, we examined the effects of 21 major anthocyanins and the extracts from 3 food supplements containing anthocyanins on the aryl hydrocarbon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service