Skip to Content
Merck
All Photos(1)

Key Documents

44526

Supelco

L-Alanine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2-Aminopropionic acid, L-α-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
Beilstein:
1720248
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

314.5 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1

InChI key

QNAYBMKLOCPYGJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lillian L Siu et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(19), 2477-2484 (2013-05-22)
The antiepidermal growth factor receptor monoclonal antibody cetuximab has improved survival in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal cancer. The addition of brivanib, a tyrosine kinase inhibitor targeting vascular endothelial growth factor receptor and fibroblast growth factor receptor, to
Florian Madura et al.
The Journal of biological chemistry, 288(26), 18766-18775 (2013-05-24)
The T-cell receptor (TCR) recognizes peptides bound to major histocompatibility molecules (MHC) and allows T-cells to interrogate the cellular proteome for internal anomalies from the cell surface. The TCR contacts both MHC and peptide in an interaction characterized by weak
J M Rhoads et al.
Journal of pediatric gastroenterology and nutrition, 9(2), 225-231 (1989-08-01)
We studied sodium-dependent uptake of L-alanine into small intestinal brush border membrane vesicles (BBMV) isolated from piglets 40 h after infection with transmissible gastroenteritis (TGE) virus. Vesicles from TGE-infected pigs and uninfected litter-mate controls showed comparable degrees of enrichment and
Mom Das et al.
Nucleic acids research, 42(6), 3943-3953 (2013-12-29)
Errors in protein synthesis due to mispairing of amino acids with tRNAs jeopardize cell viability. Several checkpoints to prevent formation of Ala- and Cys-tRNA(Pro) have been described, including the Ala-specific editing domain (INS) of most bacterial prolyl-tRNA synthetases (ProRSs) and
Rachelle S Doody et al.
The New England journal of medicine, 369(4), 341-350 (2013-07-26)
Alzheimer's disease is characterized by the presence of cortical amyloid-beta (Aβ) protein plaques, which result from the sequential action of β-secretase and γ-secretase on amyloid precursor protein. Semagacestat is a small-molecule γ-secretase inhibitor that was developed as a potential treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service