Skip to Content
Merck
All Photos(2)

Key Documents

43827

Supelco

4-Aminoantipyrine

analytical standard

Synonym(s):

4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$155.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$155.00

About This Item

Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Beilstein:
181635
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$155.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

loss

≤0.5% loss on drying

mp

105-110 °C (lit.)

application(s)

environmental
pharmaceutical

format

neat

SMILES string

CN1N(c2ccccc2)C(=O)C(N)=C1C

InChI

1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3

InChI key

RLFWWDJHLFCNIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminoantipyrine is a type of non-steroid, anti-inflammatory drug (NSAID).[1] It is a PGE2-dependent blocker and inhibitor of cyclooxygenase (COX).[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of rose like structured LaCoO3 assisted functionalized carbon nanofiber nanocomposite for efficient electrochemical detection of anti-inflammatory drug 4-aminoantipyrine.
Sakthivel M, et al.
Electrochimica Acta, 260, 571-581 (2018)
4-Aminoantipyrine reduces toxic and genotoxic effects of doxorubicin, cisplatin, and cyclophosphamide in male mice.
Berno CR, et al.
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 805, 19-24 (2016)
Xudong Yu et al.
Materials science & engineering. C, Materials for biological applications, 40, 467-471 (2014-05-27)
In this paper, a new kind of phenol-based chemsensor L2 comprised of a Schiff base and azo groups was rationally designed and synthesized. It could selectively recognize fluoride anion among tested anions such as F(-), AcO(-), H2PO4(-), Cl(-), Br(-), and
Saloua Hatmi et al.
Journal of experimental botany, 66(3), 775-787 (2014-11-12)
Environmental factors including drought stress may modulate plant immune responses and resistance to pathogens. However, the relationship between mechanisms of drought tolerance and resistance to pathogens remained unknown. In this study, the effects of drought stress on polyamine (PA) homeostasis
Ildiko Kasza et al.
PLoS genetics, 10(8), e1004514-e1004514 (2014-08-08)
Homeostatic temperature regulation is fundamental to mammalian physiology and is controlled by acute and chronic responses of local, endocrine and nervous regulators. Here, we report that loss of the heparan sulfate proteoglycan, syndecan-1, causes a profoundly depleted intradermal fat layer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service