Skip to Content
Merck
All Photos(1)

Documents

433950

Supelco

Amberlite HPR1200 Ion Exchange Resin

hydrogen form, 23-27 mesh

Synonym(s):

strong cation exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

AmberLite HPR1200 H, 23-27 mesh particle size

form

beads

Quality Level

crosslinking

8 % cross-linked

moisture

50-56%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid

particle size

23-27 mesh
550-650 μm

capacity

1.8 meq/mL by wetted bed volume

separation technique

cation exchange

Related Categories

Application

Dowex 1X4, anion-exchange resin, was used as adsorbent material in characterization of iodinated contrast media using 11B NMR and capillary electrophoresis.
Stongly acidic cation exchange resin designed for demineralization. The monodispersed particles size range provides superior flow and efficiency. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Other Notes

This resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Rustighi et al.
Journal of hazardous materials, 205-206, 10-16 (2012-01-17)
Iodinated contrast media (ICM) are persistent and ubiquitous water pollutants. Because of their high water solubility and biochemical stability, their phase-separation and recovery from the aquatic environment is very difficult. Here, borate was chosen as a complexing agent of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service