Skip to Content
Merck
All Photos(1)

Documents

43201

Supelco

(−)-Isoledene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2085196
UNSPSC Code:
85151701

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

optical activity

[α]/D -50±2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

95 °C/5 mmHg (lit.)

density

0.902 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@H]2[C@H](C3=C1CC[C@H]3C)C2(C)C

InChI

1S/C15H24/c1-9-6-8-12-14(15(12,3)4)13-10(2)5-7-11(9)13/h9-10,12,14H,5-8H2,1-4H3/t9-,10-,12-,14-/m1/s1

InChI key

NUQDPKOFUKFKFD-BGOOENEXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Isoledene belongs to the class of flavor and fragrance standards provided for the quality control of food and cosmetic products. It is one of the important components in the essential oils of leaves and fruits of green strawberry, Pinus L. Taxa, Bidens pilosa Linn. var. Radiata, etc. (−)-Isoledene is also identified as one of the key components in Urtica dioica L. plant and exhibits anti-repellent biological activity.

Application

(−)-Isoledene may be used as an analytical reference standard for the quantification of the analyte in the extract of Zingiber officinale and Urtica dioica L. using gas-chromatography coupled to mass spectrometry (GC-MS) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential oil composition of six Pinus L. taxa (Pinaceae) from Canada and their chemotaxonomy
Kilic O, et al.
Journal of agricultural science and technology, 67-73 (2014)
Chemical composition and antioxidant, antibacterial and antifungal activities of the essential oils from Bidens pilosa Linn. var. Radiata
Deba F, et al.
Food Control, 19(4), 346-352 (2008)
Newly formulated extract of Zingiber officinale as reducing agent for Silver nitrate Nanoparticals
Hazim I, et al.
Pharma Chemica, 9(5), 232-238 (2020)
Gas Chromatography?Mass Spectrometry of Methanol Extract of Urtica dioica L. from Arunachal Pradesh, India
Kalita CB, et al.
Journal of Clinical Trials and Case Reports, 1(1) (2018)
Composition of essential oil of leaves and fruits of green strawberry (Fragaria viridis Weston) growing wild in Northern Kazakhstan
Kirillov V, et al.
Journal of Applied Botany and Food Quality, 92(5) (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service