Skip to Content
Merck
All Photos(1)

Key Documents

43019

Supelco

Ginsenoside Rg6

analytical standard

Synonym(s):

(3β,6α,12β)-3,12-Dihydroxydammara-20,24-dien-6-yl 2-O-(6-deoxy-β-L-mannopyranosyl)-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O12
CAS Number:
Molecular Weight:
767.00
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

General description

Ginsenoside Rg6 (GRg6) is a rare component of the black ginseng (BG). It is a specific protopanaxatriol (PPT)-type ginsenoside which is capable of exhibiting enhanced anti-inflammatory and anti-oxidation effects compared to commercial ginsenosides.

Application

Ginsenoside Rg6 may be used as an analytical reference standard for the determination of the analyte in Panax ginseng preparations, Chinese medicine formula Sheng-Mai San (SMS), and rat plasma post the administration of SMS by chromatography based techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional mechanism of Ginsenosides on tumor growth and metastasis
Chen T, et al.
Saudi Journal of Biological Sciences, 25(5), 917-922 (2018)
Identification of multiple constituents in the traditional Chinese medicine formula Sheng-Mai San and rat plasma after oral administration by HPLC-DAD-MS/MS
Wang YH, etal.
Journal of Pharmaceutical and Biomedical Analysis, 54(5), 1110-1127 (2011)
Simultaneous determination of 30 ginsenosides in Panax ginseng preparations using ultra performance liquid chromatography
Park HW, et al.
Journal of ginseng research, 37(4), 457-457 (2013)
Rapid affinity chromatographic isolation method for LDL in human plasma by immobilized chondroitin-6-sulfate and anti-apoB-100 antibody monolithic disks in tandem
Liangsupree T, et al.
Scientific Reports, 9(1), 1-10 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service