Skip to Content
Merck
All Photos(1)

Key Documents

42739

Supelco

Cyanidin 3-sophoroside chloride

analytical standard

Synonym(s):

Cyanidin 3-O-(2-O-β-D-glucopyranosyl)-β-D-glucopyranoside chloride, Cyanidin 3-diglucoside, Sophoroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31ClO16
CAS Number:
Molecular Weight:
646.98
UNSPSC Code:
85151701

grade

analytical standard

Assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Cyanidin 3-sophoroside is classified under the family of anthocyanin flavonoids, which are ubiquitously present as pigments in fruits, flowers and vegetables. This phenolic bioactive compound is known to exhibit powerful free radical scavenging activity.[1][2][3] Cyanidin 3-sophoroside chloride is generally available as a chloride salt.

Application

Cyanidin 3-sophoroside chloride may be used as an analytical standard for the determination of the analyte in plant extracts[4] and fruits[5] by high performance liquid chromatography (HPLC) technique.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative analyses of copigmentation of cyanidin 3-glucoside and cyanidin 3-sophoroside from red raspberry fruits
Sun J, et al.
Food Chemistry, 120(4), 1131-1137 (2010)
Impact of packaging material and storage conditions on polyphenol stability, colour and sensory characteristics of freeze-dried sour cherry (prunus cerasus var. Marasca)
Zoric Z, et al.
Journal of Food Science and Technology, 53(2), 1247-1258 (2016)
Red fruit juice quality and authenticity control by HPLC
Obon JM, et al.
Journal of food composition and analysis, 24(6), 760-771 (2011)
Light and heat sensitivity of red cabbage extract in soft drink model systems
Dyrby M, et al.
Food Chemistry, 72(4), 431-437 (2001)
Phenolics, antioxidants and color properties of aqueous pigmented plant extracts: Ardisia colorata var. elliptica, Clitoria ternatea, Garcinia mangostana and Syzygium cumini
Azima AMS, et al.
Journal of functional foods, 38, 232-241 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service