Skip to Content
Merck
All Photos(1)

Documents

42697

Supelco

Diethyl adipate

analytical standard

Synonym(s):

Hexanedioic acid 1,6-diethyl ester, NSC 19160, NSC 3363

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)4COOC2H5
CAS Number:
Molecular Weight:
202.25
Beilstein:
1780035
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.427 (lit.)

bp

251 °C (lit.)

mp

−20-−19 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(=O)CCCCC(=O)OCC

InChI

1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3

InChI key

VIZORQUEIQEFRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Ion Mobility: TWCCSN2 value of 152.6 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 42697 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service