Skip to Content
Merck
All Photos(1)

Key Documents

42248

Supelco

7-Methoxycoumarin

analytical standard

Synonym(s):

Herniarin, Methyl umbelliferyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Beilstein:
141728
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

mp

117-121 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccc2C=CC(=O)Oc2c1

InChI

1S/C10H8O3/c1-12-8-4-2-7-3-5-10(11)13-9(7)6-8/h2-6H,1H3

InChI key

LIIALPBMIOVAHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-Mei Ma et al.
Phytochemical analysis : PCA, 18(1), 42-49 (2007-01-31)
A simple HPLC-PAD-MS method was established to quantitatively analyse two spiroether isomers (cis-en-yn-dicycloether and trans-en-yn-dicycloether) and the main coumarin, herniarin, in chamomile herbs, simultaneously. By using this method, the contents of these three compounds in the flowers of two chamomile
P Paterson et al.
Xenobiotica; the fate of foreign compounds in biological systems, 14(11), 849-859 (1984-11-01)
Pretreatment of rats with phenobarbitone increased hepatic microsomal 7-methoxy-and 7-ethoxy-coumarin O-dealkylase activities. Pretreatment with beta-naphthoflavone increased only the 7-ethoxycoumarin O-dealkylase activity. The addition of metyrapone in vitro inhibited the O-dealkylations to different extents. Similar results were obtained with diphenyloxazole and
M Warren et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(8), 973-981 (1988-08-01)
1. The activities of 7-methoxycoumarin (7-MCOD), 7-ethoxycoumarin (7-ECOD) and 7-propoxycoumarin (7-PCOD) O-dealkylases decreased by 75-90% during culture of rat hepatocytes for 72 h. 2. The addition of dexamethasone (D) produced a stabilization or modest enhancement of 7-ECOD and 7-PCOD activities
Adriana Eliasová et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 59(7-8), 543-548 (2005-04-09)
The responses of young plants of diploid and tetraploid Matricaria chamomilla cultivars to abiotic stress were studied. The course of quantitative changes of main leaf secondary metabolites was evaluated within an interval from 6 h before to 54 h after
M Abu Mraheil et al.
Die Pharmazie, 68(7), 541-548 (2013-08-09)
Due to the increasing prevalence of antibiotic resistance and the yet low output of the genomics-based drug discovery approach novel strategies are urgently needed to detect new antibiotics. One such strategy uses known ubiquitous targets like DNA topoisomerases. However, to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service