Skip to Content
Merck
All Photos(2)

Key Documents

42147

Supelco

(RS)-Mevalonic acid lithium salt

analytical standard

Synonym(s):

(±)-MVA-Li, rac.-MVA-Li, Lithium (±)-3,5-dihydroxy-3-methyl-pentanoate, Lithium (±)-3,5-dihydroxy-3-methyl-valerate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11O4 · xLi+
CAS Number:
Molecular Weight:
147.15 (free acid basis)
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

OC(CCO)(CC(=O)O)C

InChI

1S/C6H12O4/c1-6(10,2-3-7)4-5(8)9/h7,10H,2-4H2,1H3,(H,8,9)

InChI key

KJTLQQUUPVSXIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

NMR-Internal standard: Dimethylsulfone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andréa Hemmerlin
Plant science : an international journal of experimental plant biology, 203-204, 41-54 (2013-02-19)
Identification of regulatory enzymes is fundamental for engineering metabolic pathways such as the isoprenoid one. All too often, investigation of gene expression remains the major trend in unraveling regulation mechanisms of the isoprenoid cytosolic mevalonate and the plastid-localized methylerythritol phosphate
Martin Thurnher et al.
Biochimica et biophysica acta, 1831(6), 1009-1015 (2013-03-26)
The mevalonate pathway is a highly conserved metabolic cascade and provides isoprenoid building blocks for the biosynthesis of vital cellular products such as cholesterol or prenyl pyrophosphates that serve as substrates for the posttranslational prenylation of numerous proteins. The pathway
Ai Hosaka et al.
Neurochemical research, 38(3), 589-600 (2012-12-28)
Statins, 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase inhibitors, have been reported to attenuate amyloid-β peptide (Aβ) production in various cellular models. However, the mechanisms by which statins affect neuronal Aβ production have not yet been clarified. Here, we investigated this issue in
Adelene Ai-Lian Song et al.
PloS one, 7(12), e52444-e52444 (2013-01-10)
Isoprenoids are a large and diverse group of metabolites with interesting properties such as flavour, fragrance and therapeutic properties. They are produced via two pathways, the mevalonate pathway or the 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. While plants are the richest source of
Elena Lesma et al.
The Journal of pharmacology and experimental therapeutics, 345(2), 180-188 (2013-02-22)
Tuberous sclerosis complex (TSC) is a multi-systemic syndrome caused by mutations in TSC1 or TSC2 gene. In TSC2-null cells, Rheb, a member of the Ras family of GTPases, is constitutively activated. Statins inhibit 3-hydroxy-3-methylglutaryl coenzyme A reductase and block the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service