Skip to Content
Merck

Skip To

41003

trans-2-Hexadecenoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[[(2E)-1-oxo-2-hexadecen-1-yl]oxy]-1-propanaminium inner salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
$189.00

About This Item

Empirical Formula (Hill Notation):
C23H43NO4
CAS Number:
Molecular Weight:
397.59
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

assay

≥95.0% (LC/MS)

optical activity

[α]/D -14±2°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

[O-]C(C([H])([H])[C@](OC(/C([H])=C([H])/C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])C([H])([H])[H])=O)([H])C([H])([H])[N+](C([H])([H])[H])(C([H])([H])[H])C([H])([H])[H

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
191887677192439
format

neat

format

neat

format

neat

format

neat

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

optical activity

[α]/D -14±2°, c = 0.1 in methanol

optical activity

[α]/D -18±2°, c = 0.1 in methanol

optical activity

[α]/D -13±2°, c = 0.1 in methanol

optical activity

[α]/D -14.0±2.0°, c = 0.1 in H2O

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

assay

≥95.0% (LC/MS)

assay

≥92.0% (HPLC)

assay

≥95.0% (LC/MS)

assay

≥95.0% (HPLC)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters