Skip to Content
Merck
All Photos(1)

Key Documents

40963

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

for electrochemistry, ≥98.5% (T)

Synonym(s):

1-Butyl-1-methylpyrrolidinium N,N-bis[(trifluoromethyl)sulfonyl]amide, 1-Butyl-1-methylpyrrolidinium bis(trifluoromethanesulfonyl)amide, 1-Butyl-1-methylpyrrolidinium bistriflimide, N-Butyl-N-methylpyrrolidinium bistriflimide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$109.00

$109.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$109.00

About This Item

Empirical Formula (Hill Notation):
C11H20F6N2O4S2
CAS Number:
Molecular Weight:
422.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$109.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98.5% (T)

form

liquid

impurities

≤400 ppm water

conductivity

2.0 mS/cm

mp

-18 °C

anion traces

bromide (Br-): ≤25 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤50 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

functional group

fluoro

SMILES string

CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C9H20N.C2F6NO4S2/c1-3-4-7-10(2)8-5-6-9-10;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h3-9H2,1-2H3;/q+1;-1

InChI key

HSLXOARVFIWOQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide is an ionic liquid that can be used:
  • As a solvent for separating azeotropic mixture of 2-propanol and water.[1]
  • To study the activity of electrode materials for the electrocatalytic oxygen reduction (ORR) and oxygen evolution (OER) reactions in Mg-air batteries.[2]
  • As a lubricant for gears and machines.[3][4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of separation of water+2-propanol mixture using different ionic liquids: 1-Butyl-3-methylimidazolium bis (trifluoromethylsulfonyl) imide or 1-butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide.
Cumplido MP, et al.
The Journal of Chemical Thermodynamics, 116, 32-41 (2018)
Viscosity-pressure dependence for nanostructured ionic liquids. Experimental values for butyltrimethylammonium and 1-butyl-3-methylpyridinium bis (trifluoromethylsulfonyl) imide.
Dakkach M, et al.
The Journal of Chemical Thermodynamics, 121(7), 27-38 (2018)
Tribological Properties of Two Bis (trifluoromethylsulfonyl) imide-Based Ionic Liquids on Steel-Steel Contact.
Sanchez FL, et al.
Tribology & Lubrication Technology, 70(7), 44-44 (2014)
Electrocatalytic Oxygen Reduction and Oxygen Evolution in Mg-Free and Mg-Containing Ionic Liquid 1-Butyl-1-Methylpyrrolidinium Bis (Trifluoromethanesulfonyl) Imide.
Bozorgchenani M, et al.
ChemElectroChem, 5(18), 2600-2611 (2018)

Articles

Ionic Liquids serve as non-aqueous electrolytes in electrochemical applications, valued for their conductivity and stability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service