Skip to Content
Merck
All Photos(1)

Key Documents

407186

Sigma-Aldrich

Bromine solution

1.0 M in trimethyl phosphate

Synonym(s):

Bromide bromate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

0.51 psi ( 20 °C)
1.22 psi ( 55 °C)

concentration

1.0 M in trimethyl phosphate

density

1.307 g/mL at 25 °C (lit.)

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Muta. 1B - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shi Jun Ang et al.
Journal of computational chemistry, 40(20), 1829-1835 (2019-04-06)
Inspired by the recent interest of halogen bonding (XB) in the solid state, we detail a comprehensive benchmark study of planewave DFT geometry and interaction energy of lone-pair (LP) type and aromatic (AR) type halogen bonded complexes, using PAW and
Mohamed Edahbi et al.
Environmental science and pollution research international, 25(11), 10997-11010 (2018-02-07)
Several rare earth element (REE) mine projects around the world are currently at the feasibility stage. Unfortunately, few studies have evaluated the contamination potential of REE and their effects on the environment. In this project, the waste rocks from the
Ian Parker et al.
Scientific reports, 7(1), 11029-11029 (2017-09-10)
Membrane nanotubes are cytosolic protrusions with diameters <1 µm that extend between cells separated by tens of µm. They mediate several forms of intercellular communication and are upregulated in diverse diseases. Difficulties in visualizing and studying nanotubes within intact tissues have
Mohammed S Abdelbassit et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(50), 11659-11669 (2019-07-11)
A series of octanuclear iodine-bromine interhalides [In Br8-n ]2- (n=0, 2, 3, 4) were prepared systematically in two steps. Firstly, addition of a dihalogen (Br2 or IBr) to the triaminocyclopropenium bromide salt [C3 (NEt2 )3 ]Br forms the corresponding trihalide
Juan Liu et al.
Journal of hazardous materials, 384, 121378-121378 (2019-10-14)
Thallium (Tl) is typical rare element with severe toxicity comparable to Hg and Pb. To track Tl pollution, isotopic fractionation of Tl was evaluated during pyrite smelting for sulfuric acid production. Large variations in Tl isotope compositions were observed among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service