Skip to Content
Merck
All Photos(1)

Key Documents

40568

Millipore

Bolton Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
$165.00

$165.00


Estimated to ship onApril 14, 2025


Request more information

Select a Size

Change View
5 VIALS
$165.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

$165.00


Estimated to ship onApril 14, 2025


Request more information

Agency

according to ISO 10272-1:2017

Quality Level

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages
pathogen testing

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

This supplement is recommended for the selective enrichment of Campylobacter species from foods as recommended by ISO Committee under specifications of ISO 10272-1:2006 (E).

Application

for the selective enrichment of Campylobacter species from foods

Components

(per vial, sufficient for 500 ml medium):
Cefoperazone 10 mg
Vancomycin 10 mg
Trimethoprim 10 mg
Amphotericin 5 mg

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service