Skip to Content
Merck
All Photos(1)

Key Documents

39885

Millipore

Rose Bengal Chloramphenicol Agar acc. GB

suitable for microbiology, NutriSelect® Plus

Synonym(s):

RBC Agar, RBCA

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$44.50

$44.50


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$44.50

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

$44.50


Estimated to ship onMay 17, 2025


Request a Bulk Order

Agency

according to GB 4789.15-2016

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20 g/L
Chloramphenicol, 0.1 g/L
glucose, 10 g/L
magnesium sulfate, 0.5 g/L
peptone, 5 g/L
potassium dihydrogen phosphate, 1 g/L
Rose Bengal, 0.033 g/L

manufacturer/tradename

NutriSelect® Plus

application(s)

environmental
food and beverages

microbiology

suitability

molds
yeasts

Application

For the selective isolation and enumeration of yeasts and moulds from environmental materials and food stuffs.

Preparation Note

Suspend 36.6 g in 1 litre distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service