Skip to Content
Merck
All Photos(1)

Documents

39587

Supelco

2-Methylfuran

analytical standard

Synonym(s):

Silvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
Beilstein:
103733
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.8 (vs air)

vapor pressure

139 mmHg ( 20 °C)
561 mmHg ( 55 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.432-1.434
n20/D 1.433 (lit.)

bp

63-66 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1ccco1

InChI

1S/C5H6O/c1-5-3-2-4-6-5/h2-4H,1H3

InChI key

VQKFNUFAXTZWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylfuran may be used as a reference standard for the determination of the analyte in fruit juices by headspace solid phase microextraction (HS-SPME) procedure, followed by gas chromatography-flame ionization detection (GC-FID) .
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

An efficient method for the simultaneous determination of furan, 2-methylfuran and 2-pentylfuran in fruit juices by headspace solid phase microextraction and gas chromatography?flame ionisation detector.
Hu, Gaofei, et al.
Food Chemistry, 192, 9-14 (2016)
Maryam Chaichi et al.
Food additives & contaminants. Part B, Surveillance, 8(1), 73-80 (2014-10-31)
In this study, the levels of furan, 2-methylfuran, 2,5-dimethylfuran, vinyl furan, 2-methoxymethyl-furan and furfural in different coffee products were evaluated. Simultaneous determination of these six furanic compounds was performed by a head space liquid-phase micro-extraction (HS-LPME) method. A total of
Benjamin Sachse et al.
Carcinogenesis, 35(10), 2339-2345 (2014-07-24)
Furfuryl alcohol is a rodent carcinogen present in numerous foodstuffs. Sulfotransferases (SULTs) convert furfuryl alcohol into the DNA reactive and mutagenic 2-sulfoxymethylfuran. Sensitive techniques for the isotope-dilution ultra performance liquid chromatography-tandem mass spectrometry quantification of resulting DNA adducts, e.g. N

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service