Skip to Content
Merck
All Photos(1)

Key Documents

39484

Millipore

Methyl Red Voges Proskauer Broth

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Buffered Glucose Broth, Glucose Phosphate Broth, MR VP Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

according to GB 4789.30-2016
according to ISO 11290-1:2017
according to ISO 11290-2:2017
according to ISO 22964:2017

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bufferes peptone, 7 g/L
dextrose, 5 g/L
dipotassium phosphate, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | metabolite detection: suitable
microbiological culture: suitable

final pH

6.9±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

nonselective for Citrobacter spp.
nonselective for Escherichia coli
nonselective for Klebsiella spp.
nonselective for Proteus spp.
nonselective for Salmonella spp.
nonselective for Yersinia spp.
nonselective for coliforms
nonselective for enterobacteriaceae

Application

A classic liquid medium recommended for the Methyl-red and Voges-Proskauer tests for the differentiation of the coli-aerogenes group according ISO standards 6579 and 6585 and FIL - IDF 93 standard.

Preparation Note

Dissolve 17 g of in 1 litre of distilled water. Mix well, distribute into test tubes in amounts of 10 ml and sterilise by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beitrag zur Ernahrungsphysiologie and zur Differential diagnose der Bakterien der hamorrhagischen Septicamie.
Voges, D. and Proskauer, B.
Z. Hyg., 28, 20-32 (1898)
Notes on Bacillus coli communis and belated forms, together with some suggestions concerning the bacteriological examination of drinking-water.
Smith, T.
The American Journal of the Medical Sciences, 110, 283-301 (1895)
The differentiation of the colon-aerogenes family by the use of indicators.
Clark, W. M. and Lubs, H. A.
The Journal of Infectious Diseases, 17, 160-173 (1915)

Articles

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service