Skip to Content
Merck
All Photos(2)

Key Documents

389404

Sigma-Aldrich

5,10,15,20-Tetraphenyl-21H, 23H-porphine-p,p′,p,p′′′-tetrasulfonic acid tetrasodium hydrate

Synonym(s):

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate, meso-Tetraphenylporphine-4,4′,4″,4″′-tetrasulfonic acid tetrasodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$200.00
500 MG
$646.00

$200.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$200.00
500 MG
$646.00

About This Item

Empirical Formula (Hill Notation):
C44H26N4Na4O12S4 · xH2O
CAS Number:
Molecular Weight:
1022.92 (anhydrous basis)
Beilstein:
5488862
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$200.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

form

powder, crystals or chunks

Quality Level

λmax

414 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(cc4)S([O-])(=O)=O)c5ccc([nH]5)c(-c6ccc(cc6)S([O-])(=O)=O)c7ccc(n7)c(-c8ccc(cc8)S([O-])(=O)=O)c9ccc2[nH]9

InChI

1S/C44H30N4O12S4.4Na.H2O/c49-61(50,51)29-9-1-25(2-10-29)41-33-17-19-35(45-33)42(26-3-11-30(12-4-26)62(52,53)54)37-21-23-39(47-37)44(28-7-15-32(16-8-28)64(58,59)60)40-24-22-38(48-40)43(36-20-18-34(41)46-36)27-5-13-31(14-6-27)63(55,56)57;;;;;/h1-24,45,48H,(H,49,50,51)(H,52,53,54)(H,55,56,57)(H,58,59,60);;;;;1H2/q;4*+1;/p-4/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;;;;

InChI key

LMNVUEGSCDLLCZ-FLYAKZGESA-J

Looking for similar products? Visit Product Comparison Guide

General description

5,10,15,20-Tetraphenyl-21H, 23H-porphine-p,p′,p,p′′′-tetrasulfonic acid tetrasodium hydrate is a water-soluble porphyrin. It is a negatively charged compound due to the presence of the sulfonate groups.

Application

5,10,15,20-Tetraphenyl-21H, 23H-porphine-p,p′,p,p′′′-tetrasulfonic acid tetrasodium hydrate has been used to functionalize chemically converted graphene to form conductive graphene films of varying thickness.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porphyrin functionalized graphene sheets in aqueous suspensions: from the preparation of graphene sheets to highly conductive graphene films.
Geng J and Tung HT
The Journal of Physical Chemistry C, 114(18), 8227-8234 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service