Skip to Content
Merck
All Photos(2)

Key Documents

38464

Supelco

Disperse Yellow 9

analytical standard

Synonym(s):

N-(2,4-Dinitrophenyl)-1,4-phenylenediamine, N-(2,4-Dinitrophenyl)-p-phenylenediamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$180.00

$180.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$180.00

About This Item

Linear Formula:
(O2N)2C6H3NHC6H4NH2
CAS Number:
Molecular Weight:
274.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$180.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

187-190 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Nc1ccc(Nc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O)cc1

InChI

1S/C12H10N4O4/c13-8-1-3-9(4-2-8)14-11-6-5-10(15(17)18)7-12(11)16(19)20/h1-7,14H,13H2

InChI key

JMDHCJDATBJFJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Davide Tamburro et al.
Journal of the American Chemical Society, 133(47), 19178-19188 (2011-10-18)
Many low-abundance biomarkers for early detection of cancer and other diseases are invisible to mass spectrometry because they exist in body fluids in very low concentrations, are masked by high-abundance proteins such as albumin and immunoglobulins, and are very labile.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service