Skip to Content
Merck
All Photos(2)

Key Documents

383783

Sigma-Aldrich

N,N′-Ditridecylperylene-3,4,9,10-tetracarboxylic diimide

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$496.00

$496.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$496.00

About This Item

Empirical Formula (Hill Notation):
C50H62N2O4
CAS Number:
Molecular Weight:
755.04
Beilstein:
8607257
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$496.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder, crystals or chunks

mp

>300 °C (lit.)

solubility

toluene: 10 mg/mL

λmax

526 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCN1C(=O)c2ccc3c4ccc5C(=O)N(CCCCCCCCCCCCC)C(=O)c6ccc(c7ccc(C1=O)c2c37)c4c56

InChI

1S/C50H62N2O4/c1-3-5-7-9-11-13-15-17-19-21-23-33-51-47(53)39-29-25-35-37-27-31-41-46-42(32-28-38(44(37)46)36-26-30-40(48(51)54)45(39)43(35)36)50(56)52(49(41)55)34-24-22-20-18-16-14-12-10-8-6-4-2/h25-32H,3-24,33-34H2,1-2H3

InChI key

XCAZCFDCJHGAIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dyes and Pigments, 11, 303-303 (1989)

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service