Skip to Content
Merck
All Photos(1)

Key Documents

37923

Supelco

Imazethapyr

PESTANAL®, analytical standard

Synonym(s):

2-(4,5-Dihydro-4-isopropyl-4-methyl-5-oxo-1H-imidazol-2-yl)-5-ethyl-3-pyridinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N3O3
CAS Number:
Molecular Weight:
289.33
Beilstein:
7437150
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1cnc(C2=NC(C)(C(C)C)C(=O)N2)c(c1)C(O)=O

InChI

1S/C15H19N3O3/c1-5-9-6-10(13(19)20)11(16-7-9)12-17-14(21)15(4,18-12)8(2)3/h6-8H,5H2,1-4H3,(H,19,20)(H,17,18,21)

InChI key

XVOKUMIPKHGGTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imazethapyr is an imidazolinone derivative and a herbicide, which is selectively used on crops for weed control management.

Application

Imazethapyr may be used as a reference standard in the determination of imazethapyr in soil samples using high performance liquid chromatography coupled with ultraviolet detector (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of the herbicide imazethapyr on soil microbial biomass and various soil enzyme activities
Perucci P and Scarponi L.
Biology and Fertility of Soils, 17(3), 237-240 (1994)
Jacob S Montgomery et al.
Genome biology and evolution, 12(11), 1988-1993 (2020-08-25)
Amaranthus tuberculatus, Amaranthus hybridus, and Amaranthus palmeri are agronomically important weed species. Here, we present the most contiguous draft assemblies of these three species to date. We utilized a combination of Pacific Biosciences long-read sequencing and chromatin contact mapping information
Haifeng Qian et al.
Environmental science & technology, 45(16), 7036-7043 (2011-07-14)
Imazethapyr (IM) is a chiral herbicide and a widely used racemic mixture. This report investigated the enantioselectivity between R- and S-IM in rice and explored its causative mechanism at the physiological and molecular levels. The results suggested that R-IM inhibited
HaiFeng Qian et al.
PloS one, 6(5), e19451-e19451 (2011-05-17)
The enantiomers of a chiral compound possess different biological activities, and one of the enantiomers usually shows a higher level of toxicity. Therefore, the exploration of the causative mechanism of enantioselective toxicity is regarded as one of primary goals of
Determination of imazethapyr and imazapyr residues in soil by coupled-column liquid chromatography
Novakova O.
Chromatographia, 29(1-2), 62-66 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service