Skip to Content
Merck
All Photos(1)

Key Documents

37893

Supelco

Cinosulfuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N5O7S
CAS Number:
Molecular Weight:
413.41
Beilstein:
8162781
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

140-145 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

COCCOc1ccccc1S(=O)(=O)NC(=O)Nc2nc(OC)nc(OC)n2

InChI

1S/C15H19N5O7S/c1-24-8-9-27-10-6-4-5-7-11(10)28(22,23)20-13(21)16-12-17-14(25-2)19-15(18-12)26-3/h4-7H,8-9H2,1-3H3,(H2,16,17,18,19,20,21)

InChI key

WMLPCIHUFDKWJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>291.2 °F - closed cup

Flash Point(C)

> 144 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Koo Lee et al.
Chemosphere, 49(2), 173-181 (2002-10-12)
In order to elucidate the long-term fate of the sulfonylurea herbicide cinosulfuron, the 14C-labelled chemical was applied to a clay loam soil, encased in two lysimeters, 22 days after rice (Oryza sativa L.) transplanting, and rice plants were grown for
C Sabater et al.
Chemosphere, 46(7), 953-960 (2002-05-10)
The acute toxicity of sulfonylurea herbicides bensulfuron-methyl and cinosulfuron was tested on the five species of freshwater phytoplankton: Scenedesmus acutus, Scenedesmus subspicatus, Chlorella vulgaris and Chlorella saccharophila. Herbicide concentrations eliciting a 50% growth reduction over 96 h (EC50) ranged from
Michèle Nègre et al.
Pest management science, 61(7), 675-681 (2005-02-24)
Cinosulfuron is a sulfonylurea herbicide largely used in the extensive cultures of flooded rice in North Italy. The degradation of cinosulfuron has been investigated in sterile aqueous solutions at 30 degrees C at different pH values. It was rapidly degraded
Alfonso Infantino et al.
Chemosphere, 70(7), 1298-1308 (2007-09-04)
A dynamic aquatic model (DynA model) was previously developed to predict the fate of a chemical in aquatic scenarios characterized by daily or periodic changes in several input parameters. DynA model is here calibrated with data obtained from the literature

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service