Skip to Content
Merck
All Photos(2)

Key Documents

37052

Supelco

Desnitro-imidacloprid hydrochloride

PESTANAL®, analytical standard

Synonym(s):

1-(2-Chloro-pyridin-5-yl-methyl)-2-imino-imidazolidine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$700.00

$700.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$700.00

About This Item

Empirical Formula (Hill Notation):
C9H11ClN4 · HCl
CAS Number:
Molecular Weight:
247.12
Beilstein:
13650656
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$700.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cl.Clc1ccc(CN2CCNC2=N)cn1

InChI

1S/C9H11ClN4.ClH/c10-8-2-1-7(5-13-8)6-14-4-3-12-9(14)11;/h1-2,5H,3-4,6H2,(H2,11,12);1H

InChI key

HAFNUTBIMPVXKF-UHFFFAOYSA-N

Related Categories

General description

Desnitro-imidacloprid hydrochloride is a metabolite of imidacloprid, a selective insecticide belonging to the class of neonicotinoids, which show good activity against pest insects in agriculture.[1]
Find more information here: Neonicotinoids

Application

Desnitro-imidacloprid hydrochloride may be used as an analytical reference standard for the quantification of the analyte in honey bees and bee products[1][2] using different chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of neonicotinoid insecticides and their metabolites in honey bee and honey by liquid chromatography tandem mass spectrometry
Gbylik-Sikorska M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 990(10), 132-140 (2015)
Refined methodology for the determination of neonicotinoid pesticides and their metabolites in honey bees and bee products by liquid chromatography- tandem mass spectrometry (LC-MS/MS)
Kamel A
Journal of Agricultural and Food Chemistry, 58(10), 5926-5931 (2010)

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service