Skip to Content
Merck
All Photos(1)

Key Documents

36993

Supelco

Bifenthrin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClF3O2
CAS Number:
Molecular Weight:
422.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Cc1c(COC(=O)[C@H]2[C@@H](\C=C(/Cl)C(F)(F)F)C2(C)C)cccc1-c3ccccc3

InChI

1S/C23H22ClF3O2/c1-14-16(10-7-11-17(14)15-8-5-4-6-9-15)13-29-21(28)20-18(22(20,2)3)12-19(24)23(25,26)27/h4-12,18,20H,13H2,1-3H3/b19-12-/t18-,20-/m1/s1

InChI key

OMFRMAHOUUJSGP-JHEGMOCKSA-N

Application

Bifenthrin solution has been used as internal standard in the determination of pesticides in tomatoes by solid–liquid extraction, followed by purification and separation at low temperature, using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F

Flash Point(C)

8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Pesticide determination in tomatoes by solid?liquid extraction with purification at low temperature and gas chromatography
Pinho dPG, et al.
Food Chemistry, 121, 251-256 (2010)
Monthathip Kongmee et al.
Journal of the American Mosquito Control Association, 28(1), 20-29 (2012-04-27)
Feeding responses of Anopheles harrisoni and An. minimus were evaluated following exposure to 2 pyrethroid insecticides, bifenthrin or deltamethrin, using an excito-repellency test system in the presence and absence of live host cues. The results demonstrated that contact irritancy was
Amanda D Harwood et al.
Chemosphere, 90(3), 1117-1122 (2012-10-23)
Recent studies have determined that techniques, such as solid phase microextraction (SPME) fibers and Tenax beads, can predict bioaccumulation and potentially could predict toxicity for several compounds and species. Toxicity of bifenthrin was determined using two standard sediment toxicity tests
Xiangwei You et al.
Bulletin of environmental contamination and toxicology, 90(2), 238-241 (2012-12-06)
Field trials were carried out to investigate the dissipation and residue levels of bifenthrin in wheat. After extraction with acetonitrile, the samples were cleaned up by dispersive solid-phase extraction and detected by gas chromatography-mass spectrometry. The half-lives of bifenthrin in
Timothy P Hurst et al.
Journal of medical entomology, 49(5), 1021-1026 (2012-10-03)
The pyrethroid insecticide Biflex AquaMax (bifenthrin) was evaluated for its ability to suppress coastal mosquito populations in a controlled suburban backyard study in Mango Hill, Queensland, Australia. The insecticide was applied to perimeter vegetation, fencing in selected backyards, or both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service