Skip to Content
Merck
All Photos(1)

Key Documents

36955

Supelco

Benzo[b]fluoranthene solution

100 μg/mL in cyclohexane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1872553
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc-2c(c1)-c3cccc4c3c-2cc5ccccc45

InChI

1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H

InChI key

FTOVXSOBNPWTSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F

Flash Point(C)

-18 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nesnow et al.
Environmental health perspectives, 101 Suppl 3, 37-42 (1993-10-01)
DNA-carcinogen adducts offer a potential dosimeter for environmental genotoxicants reaching the exposed individual. Because the target tissues for many chemical carcinogens are not readily accessible for monitoring adducts in humans, peripheral blood lymphocytes (PBLs) have served as surrogate sources of
M J Mass et al.
Carcinogenesis, 17(8), 1701-1704 (1996-08-01)
The polycyclic aromatic hydrocarbon benzo[b]fluoranthene (B[b]F) is a pervasive constituent of environmental combustion products. We sought to examine the lung tumorigenic activity of B[b]F in strain A/J mice, to study the relationship between formation and decay of B[b]F-DNA adducts and
H B Zhang et al.
Environmental pollution (Barking, Essex : 1987), 141(1), 107-114 (2005-10-26)
Surface soil (0-10 cm) samples from 53 sampling sites including rural and urban areas of Hong Kong were collected and analyzed for 16 EPA priority polycyclic aromatic hydrocarbons (PAHs). Total PAH concentrations were in the range of 7.0-410 microg kg(-1)
S Aubin et al.
Journal of the Air & Waste Management Association (1995), 50(12), 2093-2101 (2001-01-05)
Benzo[b]fluoranthene (B[b]F) was used in relative abundance ratios (RARs), a parameter obtained by dividing the concentration of individual polycyclic aromatic hydrocarbons (PAHs) found in a given sample by the concentration of B[b]F in the same sample. The B[b]F RARs were
Yeh-Chung Chien et al.
Archives of toxicology, 84(10), 767-776 (2010-06-24)
Although urinary 1-hydroxypyrene (1-OHP) is the most relevant parameter for assessing exposure to polycyclic aromatic hydrocarbons, the inability to further elucidate the intra- and inter-individual variability, specificity and kinetics makes it difficult to enhance its value as an exposure predictor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service