Skip to Content
Merck
All Photos(2)

Documents

36724

Supelco

4-Ethylphenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1363317
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

4.2 (vs air)

vapor pressure

0.13 mmHg ( 20 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

218-219 °C (lit.)

mp

40-42 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCc1ccc(O)cc1

InChI

1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3

InChI key

HXDOZKJGKXYMEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Benito et al.
Journal of applied microbiology, 106(5), 1743-1751 (2009-02-20)
The formation of ethylphenols in wines, a consequence of Dekkera/Brettanomyces metabolism, can affect their quality. The main aims of this work were to further our knowledge of Dekkera/Brettanomyces with respect to ethylphenol production, and to develop a methodology for detecting
Cyrille Sabot et al.
Chemical communications (Cambridge, England), (20)(20), 2941-2943 (2009-05-14)
A concise total synthesis of (+/-)-aspidospermidine has been achieved in twelve steps from inexpensive 4-ethylphenol, based on an oxidative Hosomi-Sakurai reaction via the concept of "aromatic ring umpolung".
Isa Silva et al.
International journal of food microbiology, 145(2-3), 471-475 (2011-02-19)
This work aimed to evaluate the effect of certain factors on the production of volatile phenols from the metabolism of p-coumaric acid by lactic acid bacteria (LAB) (Lactobacillus plantarum, L. collinoides and Pediococcus pentosaceus). The studied factors were: pH, L-malic
Hajo Zech et al.
Proteomics, 11(16), 3380-3389 (2011-07-14)
Two-dimensional difference gel electrophoresis (2-D DIGE) allows for reliable quantification of global protein abundance changes. The threshold of significance for protein abundance changes depends on the experimental variation (biological and technical). This study estimates biological, technical and total variation inherent
Michael R L Stratford et al.
Bioorganic & medicinal chemistry, 20(14), 4364-4370 (2012-06-16)
In vitro studies, using combined spectrophotometry and oximetry together with hplc/ms examination of the products of tyrosinase action demonstrate that hydroquinone is not a primary substrate for the enzyme but is vicariously oxidised by a redox exchange mechanism in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service