Skip to Content
Merck
All Photos(1)

Key Documents

36627

Supelco

1,2,4-Trichlorobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$34.20

$34.20


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$34.20

About This Item

Empirical Formula (Hill Notation):
C6H3Cl3
CAS Number:
Molecular Weight:
181.45
Beilstein:
956819
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$34.20


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

>6 (vs air)

vapor pressure

1 mmHg ( 40 °C)

product line

PESTANAL®

autoignition temp.

1060 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.6 %, 150 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.571 (lit.)

bp

214 °C (lit.)

mp

16 °C (lit.)

density

1.454 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(Cl)c(Cl)c1

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4-Trichlorobenzene can be used as a solvent for industrial purposes.[1] It can also find applications as a termiticide and lubricant for household uses.[1]

Application

1,2,4-Trichlorobenzene may be used as an eluent in order to determine the polydispersity index and also the molecular weight of benzo[c][1,2,5]oxadiazole based copolymers using high-temperature gel-permeation chromatography (GPC).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

National study of chemical residues in fish Volume II (1993)
Novel benzo [c][1, 2, 5] oxadiazole-naphthalenediimide based copolymer for high-performance air-stable n-type field-effect transistors exhibiting high electron mobility of 2.43 cm 2 V- 1 s- 1
Zhao Z, et al.
Journal of Materials Chemistry, 5(11), 2892-2898 (2017)
Vijai Elango et al.
Chemosphere, 84(2), 247-253 (2011-05-03)
Using inoculum from a microcosm study that exhibited aerobic transformation of cis-1,2-dichloroethene (cDCE) and trichloroethene (TCE) commensurate with biodegradation of monoaromatic compounds, enrichment cultures were developed by providing benzene, chlorobenzene (CB), dichlorobenzene (DCB) isomers and 1,2,4-trichlorobenzene as carbon and energy
Ernest Marco-Urrea et al.
Bioresource technology, 100(23), 5757-5762 (2009-07-21)
Extracellular hydroxyl radical ((*)OH) production via quinone redox cycling in Trametes versicolor, grown in a chemically defined medium, was investigated to degrade trichloroethylene (TCE), perchloroethylene (PCE), 1,2,4- and 1,3,5-trichlorobenzene (TCB). The activity of the enzymes catalyzing the quinone redox cycle
Anke Wagner et al.
Applied and environmental microbiology, 75(7), 1876-1884 (2009-02-10)
The transcription of reductive dehalogenase homologous (rdh) genes of "Dehalococcoides" sp. strain CBDB1 was investigated during the growth and reductive dechlorination of 1,2,3- and 1,2,4-trichlorobenzene (TCB). A method was developed to monitor the expression of all 32 rdhA genes present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service