Skip to Content
Merck
All Photos(2)

Key Documents

36518

Supelco

2,3,5,6-Tetrachlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4O
CAS Number:
Molecular Weight:
231.89
Beilstein:
2049586
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1c(Cl)c(Cl)cc(Cl)c1Cl

InChI

1S/C6H2Cl4O/c7-2-1-3(8)5(10)6(11)4(2)9/h1,11H

InChI key

KEWNKZNZRIAIAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5,6-Tetrachlorophenol is a chlorinated compound, occurring as a major contaminant in soil and groundwater.[1]

Application

2,3,5,6-Tetrachlorophenol may be used as an analytical reference standard for the quantification of the analyte in aqueous samples using gas chromatography coupled to mass spectrometry (GC-MS).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Chauhan et al.
Applied microbiology and biotechnology, 52(2), 261-266 (1999-09-28)
Degradation of 2,4,5-trichlorophenol (2,4,5-TCP) and 2,3,5,6-tetrachlorophenol (TeCP) was studied using a two-stage approach that utilized efficient pulse electric discharge (PED) followed by biological degradation with a consortium from acclimated return activated sludge. The chlorinated phenols were treated in the PED
Determination of phenols and chlorophenols as trimethylsilyl derivatives using gas chromatography-mass spectrometry.
Kovacs A, et al.
Journal of Chromatography A, 1194(1), 139-142 (2008)
Acute dermal toxicity of tetrachlorophenols in the rat.
S Y Shen et al.
Bulletin of environmental contamination and toxicology, 31(6), 680-685 (1983-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service