Skip to Content
Merck
All Photos(1)

Key Documents

36157

Supelco

Methfuroxam

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15NO2
CAS Number:
Molecular Weight:
229.27
Beilstein:
6397976
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1oc(C)c(C(=O)Nc2ccccc2)c1C

InChI

1S/C14H15NO2/c1-9-10(2)17-11(3)13(9)14(16)15-12-7-5-4-6-8-12/h4-8H,1-3H3,(H,15,16)

InChI key

ZWJNEYVWPYIKMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methfuroxam may be used as an analytical reference standard for the determination of the analyte in tea,[1] food commodities,[2] grains,[3] surface and drinking water[4] samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of multiresidual phenyl acetanilide pesticides in different food commodities by solid-phase cleanup and gas chromatography-mass spectrometry.
Li Y, et al.
Journal of Separation Science, 36(6), 1061-1069 (2013)
Fungicide methfuroxan administered chronically to rats did not affect serum thyroid hormone levels.
K Kita et al.
Central European journal of public health, 3(3), 127-128 (1995-08-01)
Simultaneous determination of 103 pesticide residues in tea samples by LC-MS/MS.
Huang z, et al.
Journal of Separation Science, 32(9), 1294-1301 (2009)
Multiresidue analysis of polar pesticides in surface and drinking water by on-line enrichment and thermospray LC-MS.
Sennert S, et al
Fresenius Journal of Analytical Chemistry, 351(7), 642-649 (1995)
Simultaneous determination of 405 pesticide residues in grain by accelerated solvent extraction then gas chromatography-mass spectrometry or liquid chromatography-tandem mass spectrometry.
Pang GF, et al.
Analytical and Bioanalytical Chemistry, 384(6), 1366-1408 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service