Skip to Content
Merck
All Photos(1)

Key Documents

34364

Supelco

Norflurazon

PESTANAL®, analytical standard

Synonym(s):

4-Chloro-5-(methylamino)-2-(α,α,α-trifluoro-m-tolyl)-3(2H)-pyridazinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$226.00

$226.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$226.00

About This Item

Empirical Formula (Hill Notation):
C12H9ClF3N3O
CAS Number:
Molecular Weight:
303.67
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$226.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC1=C(Cl)C(=O)N(N=C1)c2cccc(c2)C(F)(F)F

InChI

1S/C12H9ClF3N3O/c1-17-9-6-18-19(11(20)10(9)13)8-4-2-3-7(5-8)12(14,15)16/h2-6,17H,1H3

InChI key

NVGOPFQZYCNLDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Norflurazon is a weed controlling agent which acts by inhibiting carotenoid biosynthesis.[1]

Application

Norflurazon may be used as an analytical reference standard for the determination of the analyte in plant tissue,[2] food stuffs[3] and ground water samples[4] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of National Survey of Pesticides analytes in groundwater by liquid chromatography with postcolumn reaction detection.
Miles CJ
Journal of Chromatography A, 592(1-2), 283-290 (1992)
1O2-mediated and EXECUTER-dependent retrograde plastid-to-nucleus signaling in norflurazon-treated seedlings of Arabidopsis thaliana.
Kim C and Apel K
Molecular plant, 6(5), 1580-1591 (2013)
Determination of the insecticide/acaricide formetanate in fresh fruit by reversed-phase liquid chromatography.
J F Lawrence et al.
Journal of agricultural and food chemistry, 29(4), 722-724 (1981-07-01)
Ouzna Abrous-Belbachir et al.
Journal of agricultural and food chemistry, 57(23), 11434-11440 (2009-12-03)
Norflurazon is a bleaching herbicide known to block carotenoid biosynthesis by inhibiting phytoene desaturase activity. Soybean plants were treated with norflurazon, and we examined the effects on the desaturation of lipid molecular species in leaves using ammonium [1-(14)C] oleate labeling.
Megan E Andriankaja et al.
Plant molecular biology, 85(3), 233-245 (2014-02-20)
The establishment of the photosynthetic apparatus during chloroplast development creates a high demand for iron as a redox metal. However, iron in too high quantities becomes toxic to the plant, thus plants have evolved a complex network of iron uptake

Questions

  1. What is the solubility and storage conditions for stock solutions for the product 34364, Norflurazon?

    1 answer
    1. Norflurazon is soluble in water (0.034 g/l at 25°C), ethanol (142 g/l at 25°C), and acetone (50 g/l at 25°C). For short-term storage (days to weeks), it is recommended to store the stock solution at 0-4°C, while for long-term storage (months), it is advisable to store it at -20°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service