Skip to Content
Merck
All Photos(2)

Key Documents

34149

Supelco

Metolachlor ESA sodium salt

PESTANAL®, analytical standard

Synonym(s):

2-[(2-Ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)-amino]2-oxo-ethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$329.00

$329.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$329.00

About This Item

Empirical Formula (Hill Notation):
C15H22NNaO5S
CAS Number:
Molecular Weight:
351.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$329.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

EPA 535

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].CCc1cccc(C)c1N(C(C)COC)C(=O)CS([O-])(=O)=O

InChI

1S/C15H23NO5S.Na/c1-5-13-8-6-7-11(2)15(13)16(12(3)9-21-4)14(17)10-22(18,19)20;/h6-8,12H,5,9-10H2,1-4H3,(H,18,19,20);/q;+1/p-1

InChI key

MVQVPBMXHBOZJW-UHFFFAOYSA-M

General description

Metolachlor ESA sodium salt is used for the control of broadleaf weeds and annual grasses in primarily corn, soybean, and sorghum. Active compounds of metolachlor pesticide may be converted by the target insects or plants or may be degraded in the environment to form the corresponding ethanesulfonic (ESA) acids, oxanillic (OA) acids, and other degradates. Metolachlor ESA sodium salt is one of the metabolites of metolachlor.[1]

Application

Metolachlor ESA sodium salt may be used as an analytical reference standard for the determination of the analyte in water by solid phase extraction and liquid chromatography/electrospray ionization tandem mass spectroscopy (LC/ESI MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eliška Rozmánková et al.
Aquatic toxicology (Amsterdam, Netherlands), 221, 105444-105444 (2020-02-23)
Herbicides and their metabolites are often detected in water bodies where they may cause adverse effects to non-target organisms. Their effects at environmentally relevant concentrations are often unclear, especially concerning mixtures of pesticides. This study thus investigated the impacts of
Robert A Yokley et al.
Analytical chemistry, 74(15), 3754-3759 (2002-08-15)
A Good Laboratory Practices (GLP) validated, multiresidue analytical method is presented for the determination of the chloroacetanilide herbicides metolachlor, acetochlor, and alachlor, the chloroacetamide herbicide dimethenamid, and their respective ethanesulfonic (ESA) and oxanillic (OA) acid degradates in ground and surface

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service