Skip to Content
Merck
All Photos(2)

Key Documents

34021

Supelco

4,4′-DDT-d8

PESTANAL®, analytical standard

Synonym(s):

1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane-d8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14HCl5D8
Molecular Weight:
362.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

mass shift

M+8

SMILES string

[2H]c1c([2H])c(c([2H])c([2H])c1Cl)C(c2c([2H])c([2H])c(Cl)c([2H])c2[2H])C(Cl)(Cl)Cl

InChI

1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

YVGGHNCTFXOJCH-PGRXLJNUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4,4′-DDT-d8 is a stable labeled isotope of the pesticide, 4,4′-DDT.[1][2]

Application

4,4′-DDT-d8 may be used as a recovery surrogate standard for the quantification of 4,4′-DDT in marine pore water by equilibrium solid-phase microextraction (SPME) followed by automated thermal desorption-gas chromatography/mass spectrometry (TD-GC-MS) using disposable optical fiber.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Extending the use of USEPA method 1625 for the analysis of 4, 4?-DDT, 4, 4?-DDD, and 4, 4?-DDE.
Behzadi H and LalancetteAR
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 44(2), 122-129 (1991)
Robert P Eganhouse et al.
Journal of chromatography. A, 1415, 38-47 (2015-09-09)
A method is described for determination of ten DDT-related compounds in marine pore water based on equilibrium solid-phase microextraction (SPME) using commercial polydimethylsiloxane-coated optical fiber with analysis by automated thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS). Thermally cleaned fiber was directly exposed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service