Skip to Content
Merck
All Photos(2)

Key Documents

34017

Supelco

Isoproturon-d6

PESTANAL®, analytical standard

Synonym(s):

3-(4-Isopropylphenyl)-1,1-dimethylurea-d6

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$219.00

$219.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$219.00

About This Item

Empirical Formula (Hill Notation):
C12H12D6N2O
CAS Number:
Molecular Weight:
212.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$219.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+6

SMILES string

[2H]C([2H])([2H])N(C(=O)Nc1ccc(cc1)C(C)C)C([2H])([2H])[2H]

InChI

1S/C12H18N2O/c1-9(2)10-5-7-11(8-6-10)13-12(15)14(3)4/h5-9H,1-4H3,(H,13,15)/i3D3,4D3

InChI key

PUIYMUZLKQOUOZ-LIJFRPJRSA-N

Related Categories

General description

Isoproturon-d6 is an isotopically labeled standard of the phenylurea herbicide isoproturon,[1][2] which is used for the pre- or post-emergence control of weeds in fruits, cotton and other crops.[3]

Application

Isoproturon-d6 may be used as internal standard in the analysis of pesticides in fine airborne particulate matter, using liquid extraction under pressure and liquid chromatographic–tandem mass spectrometric method (HPLC-MS-MS).[4]
Isoproturon-d6 may be used as an internal standard for the quantification of isoproturon in:
  • Soil samples using pressurized liquid extraction (PLE) followed by liquid chromatography coupled to mass chromatography (LC-MS) and electrospray ionization-tandem mass spectrometry (LC-ESI+-MS/MS).[1]
  • Surface water samples using online solid phase extraction–liquid chromatography–tandem mass spectrometry (SPE–LC–MS/MS).[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Risk assessment of pesticides detected in surface water of the Alqueva reservoir (Guadiana basin, southern of Portugal).
Palma P, et al.
The Science of the Total Environment, 488(1-2), 208-219 (2014)
Determination of chloroacetanilides, triazines and phenylureas and some of their metabolites in soils by pressurised liquid extraction, GC-MS/MS, LC-MS and LC-MS/MS.
Dagnac T, et al.
Journal of Chromatography A, 1067(1-2), 225-233 (2005)
Frank David et al.
Talanta, 165, 201-210 (2017-02-06)
Two low-pressure columns (Bio-Beads SX-3) and three high-pressure GPC columns were compared for clean-up of a wide range of pesticides in fatty matrices of vegetable or animal origin. The GPC fractions were analyzed by GC-MS/MS and LC-MS/MS without additional clean-up.
Aurélie Berthet et al.
Archives of toxicology, 91(11), 3529-3542 (2017-05-05)
Methylchloroisothiazolinone (MCI) and methylisothiazolinone (MI) are biocides used in many types of products such as cosmetics, paints, and cleaning agents. Skin contact is often encountered when using these products. Although MCI and MI are strong allergens and cause skin irritation
Elvio D Amato et al.
Chemosphere, 236, 124400-124400 (2019-09-24)
Recently, a new sampling device combining active and passive sampling (APS) was developed for the measurement of time-averaged concentrations of metal species and both polar and non-polar organic contaminants in water. By coupling a diffusion cell (loaded with a set

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service