Skip to Content
Merck
All Photos(1)

Key Documents

33994

Supelco

Altrenogest

VETRANAL®, analytical standard

Synonym(s):

17-α-Allyl-17-β-hydroxyoestra-4,9,11-trien-3-one, Allyltrenbolone, Regumate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CCC3=C1C=C[C@@]4(C)[C@@]2([H])CC[C@@]4(O)CC=C

InChI

1S/C21H26O2/c1-3-10-21(23)12-9-19-18-6-4-14-13-15(22)5-7-16(14)17(18)8-11-20(19,21)2/h3,8,11,13,18-19,23H,1,4-7,9-10,12H2,2H3/t18-,19+,20+,21+/m1/s1

InChI key

VWAUPFMBXBWEQY-ANULTFPQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Altrenogest can be called as a steroid or a progestational drug, which finds applications in breeding. It can mainly be used to control or suppress the condition of estrus in horses.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 1 - Repr. 1B - STOT RE 2 Oral

Target Organs

Immune system,Adrenal gland

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Neuhauser et al.
Equine veterinary journal, 41(6), 572-577 (2009-10-07)
Mares with compromised pregnancies are often treated with altrenogest to prevent abortion. However, there is only limited information about effects on the foal when altrenogest treatment is continued during final maturation of the fetus. To determine effects of altrenogest treatment
C Willmann et al.
Theriogenology, 76(1), 61-73 (2011-03-15)
A positive influence of altrenogest treatment on a retarded development of the conceptus around the beginning of placentation in mares older than 8 years could be recently demonstrated. In the present study, effects of altrenogest treatment in early-pregnant mares on
Conrad Willmann et al.
Theriogenology, 75(3), 421-428 (2010-12-15)
The treatment of early pregnant mares with a history of repeated early embryonic loss with the progestin altrenogest has become routine; however no controlled studies on the efficiency of altrenogest to prevent embryonic losses are available so far. In the
Current Therapy in Equine Medicine (2009)
Detection of altrenogest and its metabolites in post administration horse urine using liquid chromatography tandem mass spectrometry?increased sensitivity by chemical derivatisation of the glucuronic acid conjugate
Lampinen-Salomonsson M, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 833, 245-256 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service