Skip to Content
Merck
All Photos(1)

Key Documents

33985

Supelco

Propoxycarbazone sodium salt

PESTANAL®, analytical standard

Synonym(s):

2-[(4-Methyl-5-oxo-3-propoxy-1,2,4-triazolin-1-yl)carbamidosulfonyl]benzoic acid methyl ester sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N4NaO7S
CAS Number:
Molecular Weight:
420.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCOC1=NN(C(=O)N1C)C(=O)N([Na])S(=O)(=O)c2ccccc2C(=O)OC

InChI

1S/C15H18N4O7S.Na/c1-4-9-26-14-16-19(15(22)18(14)2)13(21)17-27(23,24)11-8-6-5-7-10(11)12(20)25-3;/h5-8H,4,9H2,1-3H3,(H,17,21);/q;+1/p-1

InChI key

JRQGDDUXDKCWRF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Propoxycarbazone sodium salt is a selective herbicide, used for the control of downy brome in small grains.[1]

Application

Propoxycarbazone sodium salt may be used as an analytical reference standard for the determination of the analyte in wastewater samples using ion pair centrifugal partition extraction followed by high-performance liquid chromatography with ultraviolet detection (HPLC-UV).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Poaceae Research and Application, 285, 596-604 (2012)
Removal of pesticides from wastewater by ion pair Centrifugal Partition Extraction using betaine-derived ionic liquids as extractants.
Gaetano DY, et al.
Chemical Engineering Journal, 285, 596-604 (2016)
Necola Guerrina et al.
American journal of physiology. Lung cellular and molecular physiology, 320(3), L339-L355 (2020-11-26)
The aryl hydrocarbon receptor (AhR) is a ligand-activated transcription factor whose physiological function is poorly understood. The AhR is highly expressed in barrier organs such as the skin, intestine, and lung. The lungs are continuously exposed to environmental pollutants such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service