Skip to Content
Merck
All Photos(1)

Documents

33894

Supelco

Clotrimazole

VETRANAL®, analytical standard

Synonym(s):

1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

fungi

application(s)

agriculture
environmental

format

neat

Mode of action

protein synthesis | interferes

SMILES string

Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4ccnc4

InChI

1S/C22H17ClN2/c23-21-14-8-7-13-20(21)22(25-16-15-24-17-25,18-9-3-1-4-10-18)19-11-5-2-6-12-19/h1-17H

InChI key

VNFPBHJOKIVQEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clotrimazole, a non-toxic, systemic, imidazole derivative with broad-spectrum antimycotic activity, is primarily used in the treatment of oropharyngeal candidiasis.
Its mode of action involves the inhibition of ergosterol synthesis, which leads to structural and functional damage of the cell membrane.

Application

Clotrimazole may be used as a reference standard for the determination of the analyte in topical creams and ear drops by reversed-phase high-performance liquid chromatographic (RP-HPLC) method.
Clotrimazole may be used as an analytical reference standard for the determination of the analyte in:
  • Milk samples using solid-phase extraction followed by high-performance liquid chromatography (SPE-HPLC).
  • Environmental water samples using dispersive liquid-liquid microextraction followed by high-performance liquid chromatography with tandem mass spectrometry (HPLC-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mode of action of clotrimazole.
Iwata K, et al.
Sabouraudia, 11(2), 158-166 (1973)
Clotrimazole is effective for the regression of endometriotic implants in a Wistar rat experimental model of endometriosis.
Machado D, et al.
Molecular and Cellular Endocrinology, 476, 17-26 (2018)
SPE-HPLC method for determination of ketoconazole and clotrimazole residues in cow's milk.
Beigi KAF, et al.
Journal of the Brazilian Chemical Society, 22(9), 1679-1685 (2011)
Clotrimazole treatment for prevention of oral candidiasis in patients with acute leukemia undergoing chemotherapy: Results of a double-blind study.
Cuttner J, et al.
The American Journal of Medicine, 81(5), 771-774 (1986)
Determination of beclomethasone dipropionate, clotrimazole, chloramphenicol and lidocaine in pharmaceutical formulations using a novel RP-HPLC method
Reddy MU, et al.
International Journal of Pharmacy and Biological Sciences, 2(1), 453-462 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service